Msc Flwcntrlr Gas Mass 100Sccm F/ Use W/ Corrosive Gases 1 * 1 items
MFLX41002-38
Msc Flwcntrlr Gas Mass 100Sccm F/ Use W/ Corrosive Gases 1 * 1 items
MFLX41002-38
Frequently Bought Together
Multi-position magnetic stirrers, MULTISTIRRER series
: VELP SCIENTIFIC
Magnetic hotplate stirrers, ARE/AREX 5 series
: VELP SCIENTIFIC
VWR®, Accessories for Microplate Shakers and Orbital/ Mini Shakers
: VWR Collection
Analytical sieve shaker AS 400 control
: Retsch GmbH
Accessories for laboratory mixers
: WARING BLENDOR
TOOTHED BELT 1 * 1 items
: FRITSCH
Tissue grinders, Pellet Pestle®, KIMBLE®
: DWK Life Sciences
Sample splitters, RT 6,5 - RT 75
: Retsch GmbH
Accessories for ultrasonic homogenisers, SONOPULS
: BANDELIN-ELECTRONIC
VWR®, Tissue Grinders, Potter-Elvehjem Type
: VWR Collection
FRAME SM 3 STOREY 1 * 1 items
: Edmund Bühler GmbH
Stirring shafts, moon shaped, BOLA
: Bohlender