Micropump MI0036 Service Kit for 74004-31; 1/Ea
Catalog # MFLX74005-31
Supplier: Avantor Fluid Handling
About this item
Extend the life of your gear pump head with these service kits.
- Optimize performance of your Masterflex pumps
Easy to install, these service kits avoid costly downtime of your gear pump head.
- Description:SERVICE KIT FOR 74004-31
- Environmentally Preferable:
- Warranty:1 year
- Pk:1
Frequently Bought Together