Order Entry
Italy
ContactUsLinkComponent
DISPENSER FOR PARAFILM® M For dust-free storing and convenient cutting of Parafilm® M. Made of transparent acrylic - with hinged lid - with holding device for taking one roll Parafilm® M of 10 cm width or two rolls of each 5 cm width - efficient cutt 1 * 1 items
Catalog # MARI5838900
Supplier:  MARIENFELD
DISPENSER FOR PARAFILM® M For dust-free storing and convenient cutting of Parafilm® M. Made of transparent acrylic - with hinged lid - with holding device for taking one roll Parafilm® M of 10 cm width or two rolls of each 5 cm width - efficient cutt 1 * 1 items
Catalog # MARI5838900
Supplier:  MARIENFELD
Frequently Bought Together