Cadmium acetate, anhydrous, 99+% 1 * 100 g
Catalog # 47396.22
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Aniline, EMSURE® for analysis, Supelco®
Supplier: Merck
Ethyl acetate ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 40…80 °C, EMPLURA®, Supelco®
Supplier: Merck
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Toluene ≥99%, GPR RECTAPUR® for pathology
Supplier: VWR Chemicals