RUBIADIN-1 METHYL ETHER 1 * 25 mg
Catalog # CAYM34940-25
Supplier: Cayman Chemical
Frequently Bought Together
Alcol iso-amilico, TECHNICAL
Supplier: VWR Chemicals
Etanolo 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Anisolo 99%, per analisi
Supplier: Honeywell Chemicals
N,N-Dimetilacetammide ≥99%
Supplier: Thermo Fisher Scientific
N,N-Dimetilformammide ≥99.9%, CHROMASOLV™ Plus per HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cicloesano 99.8% per HPLC
Supplier: Thermo Fisher Scientific
ter-Butil metil etere ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
1-Propanolo ≥99.9%, CHROMASOLV™ per HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Etanolammina ACS
Supplier: Thermo Fisher Scientific
Diisopropile ossido ≥98.5% (by GC) stabilizzato, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Supplier: VWR Chemicals