1-(3-Chlorophenyl)biguanide (hydrochloride) 1 * 25 g
Catalog # CAYM34726-25
Supplier: Cayman Chemical
1-(3-Chlorophenyl)biguanide (hydrochloride) 1 * 25 g
Catalog # CAYM34726-25
Supplier: Cayman Chemical
Frequently Bought Together
Acetonitrile ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Toluene ≥99%, GPR RECTAPUR® for pathology
Supplier: VWR Chemicals
Acetonitrile for DNA synthesis
Supplier: Merck
(R)-(+)-Pulegone for synthesis, Sigma-Aldrich®
Supplier: Merck
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Glycerol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 1.000 mL
Supplier: PanReac AppliChem
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99%, extra pure
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem