2-(2-Aminoethyl)-3-(4-chlorophenyl)-3-hydroxyisoindol-1-one (hydrochloride) 1 * 5 mg
Catalog # CAYM34667-5
Supplier: Cayman Chemical
2-(2-Aminoethyl)-3-(4-chlorophenyl)-3-hydroxyisoindol-1-one (hydrochloride) 1 * 5 mg
Catalog # CAYM34667-5
Supplier: Cayman Chemical
Frequently Bought Together
Petroleum spirit, 80…110 °C, TECHNICAL
Supplier: VWR Chemicals
Toluene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Cyclohexane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Triethylamine for HPLC
Supplier: Thermo Fisher Scientific
3,3'-Diaminobenzidine tetrahydrochloride dihydrate, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals