5-Bromo-4-chloro-3-indolyl--D-Glucuronide (cyclohexylammonium salt hydrate) 1 * 50 mg
Catalog # CAYM33591-50
Supplier: Cayman Chemical
5-Bromo-4-chloro-3-indolyl--D-Glucuronide (cyclohexylammonium salt hydrate) 1 * 50 mg
Catalog # CAYM33591-50
Supplier: Cayman Chemical
Frequently Bought Together
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Aeroflow tap, na, general spare part 1 * 1 items
Supplier: Avantor
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Dichloromethane 99.9% stabilised, AcroSeal® for biochemistry
Supplier: Thermo Fisher Scientific
1,10-Phenanthroline monohydrate, Supelco®
Supplier: Merck
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
1-Propanol ACS
Supplier: Thermo Fisher Scientific