5-Bromo-4-chloro-3-indolyl--D-Glucuronide (cyclohexylammonium salt hydrate) 1 * 10 mg
Catalog # CAYM33591-10
Supplier: Cayman Chemical
5-Bromo-4-chloro-3-indolyl--D-Glucuronide (cyclohexylammonium salt hydrate) 1 * 10 mg
Catalog # CAYM33591-10
Supplier: Cayman Chemical
Frequently Bought Together
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent, VOLUSOL®
Supplier: VWR Chemicals
Methanol 99.8% for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetramethylammonium hydroxide 25% (w/w) in methanol
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
m-Xylene ≥99.0% (by GC)
Supplier: TCI
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific