(2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol (Diol Derivative) 1 * 25 mg
PROOMM0043.01-0025
(2RS)-3-(Naphthalen-1-yloxy)propane-1,2-diol (Diol Derivative) 1 * 25 mg
PROOMM0043.01-0025
Frequently Bought Together
pH buffer solutions
: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo
pH buffers, ready to use
: WTW
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, DuraCal
: HAMILTON BONADUZ