Order Entry
Italy
ContactUsLinkComponent
Mix-n-Stain and trade Antibody Labeling Kits allow you to label your antibody in 30 minutes without purification. Labeling tolerates common buffers and antibody stabilizers. CF and reg 583R is a red fluorescent dye with Ex/Em at 586/609 nm. One lab 1 * 1 KIT
Catalog # 92455.
Supplier:  Biotium
Mix-n-Stain and trade Antibody Labeling Kits allow you to label your antibody in 30 minutes without purification. Labeling tolerates common buffers and antibody stabilizers. CF and reg 583R is a red fluorescent dye with Ex/Em at 586/609 nm. One lab 1 * 1 KIT
Catalog # 92455.
Supplier:  Biotium
Frequently Bought Together