Order Entry
Italy
ContactUsLinkComponent
Formal Name a-[2-(ditetradecylamino)-2-oxoethyl]-?-methoxy-poly(oxy-1,2- ethanediyl) Purity =95% Formulation A solid Solubility DMF: 10 mg/ml DMF:PBS (pH 7.2) (1:2): 0.3 mg/ml DMSO: Slightly Soluble Ethanol: 1 mg/ml 1 * 25 mg
Catalog # CAYM34336-25
Supplier:  Cayman Chemical
Formal Name a-[2-(ditetradecylamino)-2-oxoethyl]-?-methoxy-poly(oxy-1,2- ethanediyl) Purity =95% Formulation A solid Solubility DMF: 10 mg/ml DMF:PBS (pH 7.2) (1:2): 0.3 mg/ml DMSO: Slightly Soluble Ethanol: 1 mg/ml 1 * 25 mg
Catalog # CAYM34336-25
Supplier:  Cayman Chemical
Frequently Bought Together