WTB35 front case 1 * 1 items
Catalog # MEMMB50191
Supplier: MEMMERT
WTB35 front case 1 * 1 items
Product Details & Documents
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
#: 1.00030.2500
Supplier: Merck
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
#: 1.09439.1000
Supplier: Merck



