Order Entry
Italy
Orders LinkContactUsLinkComponent
Cryogenic Lamination Tapes

Cryogenic Lamination Tapes

Cryogenic lamination tape, core size: 76,2 mm, Transparent, 60 m, 50,8 mm
Catalog # GAINCLAM-50C3-200
Supplier:   GA
Cryogenic Lamination Tapes
Catalog # GAINCLAM-50C3-200
Supplier:  GA
Cryogenic Lamination Tapes
Cryogenic Lamination Tapes

Cryogenic lamination tape, core size: 76,2 mm, Transparent, 60 m, 50,8 mm

Price
53,10 €
Log in for availability
1 * 1 Roll
Product Details & Documents

Thin cryo lamination tape provides added protection against abrasion, harsh chemicals, and extreme temperatures. Designed to withstand long-term storage in liquid nitrogen and ultra-low temperature freezers, multiple freeze-thaw cycles, and transport on dry ice. This clear tape can be applied over existing labels.

  • Temperature range: −196 to +93 °C
  • Clear lamination tape protects material from extreme cryogenic conditions, including liquid phase and vapour phase liquid nitrogen
  • Resists harsh laboratory environments, including multiple freeze-thaw cycles
  • Offers protection against chemicals and solvents
  • Resistant to gamma irradiation (tested up to 50 kGy)

This ultra-thin cryo laminating tape provides excellent protection against most chemicals and reagents. It has also been designed to withstand long-term storage in liquid phase and vapor phase liquid nitrogen and ultra-low temperature freezers (as low as –196 °C) when applied on cryo vials and tubes. Resistant to dry ice, boiling water, multiple freeze-thaw cycles, and other harsh laboratory environments.

Specifications
  • Width:
    50,8 mm
  • Colour:
    Transparent
  • Description:
    Cryogenic lamination tape, core size: 76,2 mm
  • Length:
    60 m
  • Seq:
    03