Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, Septa ND11

VWR®, Septa ND11

Red PTFE/white silicone/red PTFE, 45° shore A, 1,0 mm
Codice catalogo 548-0394A
Nome del fornitore:   VWR Collection
VWR®, Septa ND11
Codice catalogo 548-0394A
Nome del fornitore:  VWR Collection
VWR®, Septa ND11
VWR®, Septa ND11

Red PTFE/white silicone/red PTFE, 45° shore A, 1,0 mm

Price
394,00 €
Log in for availability
1 * 1.000 pezzi
Product Details & Documents

Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.

  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available

Septa choice depends on the application.

Specifications
  • Hardness:
    45° shore A
  • Septum:
    Red PTFE/white silicone/red PTFE
  • Seq:
    02
  • Thickness:
    1,0 mm