Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, Septa ND11

VWR®, Septa ND11

White silicone/red PTFE, 45° shore A, 1,3 mm
Codice catalogo 548-0393A
Nome del fornitore:   VWR Collection
VWR®, Septa ND11
Codice catalogo 548-0393A
Nome del fornitore:  VWR Collection
VWR®, Septa ND11
VWR®, Septa ND11

White silicone/red PTFE, 45° shore A, 1,3 mm

Price
322,00 €
Log in for availability
1 * 1.000 pezzi
Product Details & Documents

Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.

  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available

Septa choice depends on the application.

Specifications
  • Hardness:
    45° shore A
  • Septum:
    White silicone/red PTFE
  • Seq:
    01
  • Thickness:
    1,3 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.