Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, Septa GL 45

VWR®, Septa GL 45

43 mm, Beige silicone/white PTFE, 50° shore A, 3,2 mm
Codice catalogo 548-0662A
Nome del fornitore:   VWR Collection
VWR®, Septa GL 45
Codice catalogo 548-0662A
Nome del fornitore:  VWR Collection
VWR®, Septa GL 45
VWR®, Septa GL 45

43 mm, Beige silicone/white PTFE, 50° shore A, 3,2 mm

Price
226,00 €
Log in for availability
1 * 100 pezzi
Specifications
  • Ø:
    43 mm
  • Hardness:
    50° shore A
  • Septum:
    Beige silicone/white PTFE
  • Seq:
    01
  • Thickness:
    3,2 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.