Order Entry
Italy
ContactUsLinkComponent
Inner Door Kit for MDF-DU900V-PE / MDF-DU901VHL-PE. Comprises of 2 inner doors to replace one of the two original single inner door. Purchase 2 of this item if both of the original inner doors are to be reconfigured 1 * 2 items
Catalog # SANYMDF-9ID-PW
Supplier:  PHC Corporation
Inner Door Kit for MDF-DU900V-PE / MDF-DU901VHL-PE. Comprises of 2 inner doors to replace one of the two original single inner door. Purchase 2 of this item if both of the original inner doors are to be reconfigured 1 * 2 items
Catalog # SANYMDF-9ID-PW
Supplier:  PHC Corporation
Frequently Bought Together