Order Entry
Italy
ContactUsLinkComponent
N 20 bimetal crimp cap, blue/silver, center hole Silicone blue transparent/centrical PTFE-lamination colorless Hardness: 45° shore A Thickness: 3 mm pack of 100 1 * 100 items
Catalog # MANA702341
Supplier:  MACHEREY-NAGEL
N 20 bimetal crimp cap, blue/silver, center hole Silicone blue transparent/centrical PTFE-lamination colorless Hardness: 45° shore A Thickness: 3 mm pack of 100 1 * 100 items
Catalog # MANA702341
Supplier:  MACHEREY-NAGEL
Frequently Bought Together