Order Entry
Italy
ContactUsLinkComponent
Volumetric flask, 4000 ml, class A, conformity certified, ST 34/35, PE-stopper, Diffico blue, made from DURAN tubing, 1 * 1 items
Catalog # WITG3.664.400BL
Supplier:  witeg Labortechnik
Volumetric flask, 4000 ml, class A, conformity certified, ST 34/35, PE-stopper, Diffico blue, made from DURAN tubing, 1 * 1 items
Catalog # WITG3.664.400BL
Supplier:  witeg Labortechnik
Frequently Bought Together