Reagecon 8000 µS/cm Conductivity Standard at 25°C 1 * 500 mL
Catalog # REGECSKC8M
Supplier: REAGECON
Reagecon 8000 µS/cm Conductivity Standard at 25°C 1 * 500 mL
Catalog # REGECSKC8M
Supplier: REAGECON
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
pH buffer solutions, referenced at 20 °C, traceable to SRM from NIST, Fluka™
Supplier: Honeywell Chemicals
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
Supelco® TraceCERT® Organic Reference Standard, Tebutam
Supplier: Merck