Order Entry
Italy
Orders LinkContactUsLinkComponent
Versatile for numerous temperature applications: The kit consists of an air probe, an immersion/penetration probe and a surface probe. All probes are equipped with a thermocouple (type K, class 1) and are therefore compatible with the Smart Probe tes 1 * 1 items
Versatile for numerous temperature applications: The kit consists of an air probe, an immersion/penetration probe and a surface probe. All probes are equipped with a thermocouple (type K, class 1) and are therefore compatible with the Smart Probe tes 1 * 1 items
Catalog # TEST0602.5093
Supplier:   TESTO
Versatile for numerous temperature applications: The kit consists of an air probe, an immersion/penetration probe and a surface probe. All probes are equipped with a thermocouple (type K, class 1) and are therefore compatible with the Smart Probe tes 1 * 1 items
Catalog # TEST0602.5093
Supplier:  TESTO
Versatile for numerous temperature applications: The kit consists of an air probe, an immersion/penetration probe and a surface probe. All probes are equipped with a thermocouple (type K, class 1) and are therefore compatible with the Smart Probe tes 1 * 1 items
Price
130,00 €
Log in for availability
1 * 1 pezzi