Order Entry
Italy
ContactUsLinkComponent
The testo 915i temperature kit with temperature probes will enable you to tackle your temperature measurements with greater speed, flexibility and ease: Included is a wireless thermometer testo 915i, an air probe, an immersion/penetration probe and a 1 * 1 items
Catalog # TEST0563.5915
Supplier:  TESTO
The testo 915i temperature kit with temperature probes will enable you to tackle your temperature measurements with greater speed, flexibility and ease: Included is a wireless thermometer testo 915i, an air probe, an immersion/penetration probe and a 1 * 1 items
Catalog # TEST0563.5915
Supplier:  TESTO
Frequently Bought Together