Drigalski spatulas
195 mm, 40 mm
Catalog # 231-0652
Supplier: RSG
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
- For bacteriological applications
- Can also be used as stirrer
- For even spreading or better distribution across media
Specifications
- Length:195 mm
- Blade width:40 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



