Order Entry
Italy
Orders LinkContactUsLinkComponent
3-Mercaptopropionic acid 99%

3-Mercaptopropionic acid 99%

Bottle, 500 g
Codice catalogo APOSOR52782-500G
Nome del fornitore:   Apollo Scientific
3-Mercaptopropionic acid 99%
Codice catalogo APOSOR52782-500G
Nome del fornitore:  Apollo Scientific
Numero CAS:  107-96-0
3-Mercaptopropionic acid 99%

Bottle, 500 g

Numero CAS:  107-96-0
Price
57,90 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:107-96-0
  • Formula:C₃H₆O₂S
  • MDL Number:MFCD00004897
  • Molecular Weight:106.15 g/mol
  • Boiling Pt:110…111 °C/15mmHg
  • Melting Pt:17…19 °C
  • Density:1.218 g/cm³ (25 °C)
  • Flash Pt:124 °C
  • Storage Temperature:Ambient
  • UN:2922
  • ADR:8+6.1,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.