7-Hydroxyflavone 95%
Bottle, 5 g
APOSOR1155-5G
: 7-Hydroxy-2-phenylchromen-4-one, 7-Hydroxy-2-phenyl-4-benzopyrone, 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Hydroxy-2-phenyl-4H-chromen-4-one
- :6665-86-7
- :C₁₅H₁₀O₃
- :MFCD00006835
- :238.24 g/mol
- :243…247 °C
- :Ambient
- Pack type:Bottle
- Pk:5 g