Order Entry
Italy
Orders LinkContactUsLinkComponent
 

7-Hydroxyflavone 95%

Bottle, 25 g
Catalog # APOSOR1155-25G
Irritant
Supplier:   Apollo Scientific
7-Hydroxyflavone 95%
Catalog # APOSOR1155-25G
Supplier:  Apollo Scientific
CAS Number:  6665-86-7
7-Hydroxyflavone 95%

Bottle, 25 g

CAS Number:  6665-86-7
Price
371,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  7-Hydroxy-2-phenylchromen-4-one, 7-Hydroxy-2-phenyl-4-benzopyrone, 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Hydroxy-2-phenyl-4H-chromen-4-one
  • CAS Number:6665-86-7
  • Formula:C₁₅H₁₀O₃
  • MDL Number:MFCD00006835
  • Molecular Weight:238.24 g/mol
  • Melting Pt:243…247 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications