Order Entry
Italy
Orders LinkContactUsLinkComponent
7-Hydroxyflavone 95%

7-Hydroxyflavone 95%

Bottle, 1 g
Catalog # APOSOR1155-1G
Irritant
Supplier:   Apollo Scientific
7-Hydroxyflavone 95%
Catalog # APOSOR1155-1G
Supplier:  Apollo Scientific
CAS Number:  6665-86-7
7-Hydroxyflavone 95%

Bottle, 1 g

CAS Number:  6665-86-7
Price
31,90 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  7-Hydroxy-2-phenylchromen-4-one, 7-Hydroxy-2-phenyl-4-benzopyrone, 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Hydroxy-2-phenyl-4H-chromen-4-one
  • CAS Number:6665-86-7
  • Formula:C₁₅H₁₀O₃
  • MDL Number:MFCD00006835
  • Molecular Weight:238.24 g/mol
  • Melting Pt:243…247 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.