Order Entry
Italy
Orders LinkContactUsLinkComponent
4-Methoxy-1-naphthaldehyde 95%

4-Methoxy-1-naphthaldehyde 95%

Bottle, 25 g
Codice catalogo APOSOR480638-25G
Nome del fornitore:   Apollo Scientific
4-Methoxy-1-naphthaldehyde 95%
Codice catalogo APOSOR480638-25G
Nome del fornitore:  Apollo Scientific
Numero CAS:  15971-29-6
4-Methoxy-1-naphthaldehyde 95%

Bottle, 25 g

Numero CAS:  15971-29-6
Price
57,90 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  4-Methoxynaphthalene-1-carbaldehyde
  • CAS Number:15971-29-6
  • Formula:C₁₂H₁₀O₂
  • MDL Number:MFCD00004006
  • Molecular Weight:186.21 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.