Order Entry
Italy
Orders LinkContactUsLinkComponent
 

3-Hydroxy-4-methyl-benzaldehyde 95%

Bottle, 250 mg
Catalog # APOSOR942429-250MG
Supplier:   Apollo Scientific
3-Hydroxy-4-methyl-benzaldehyde 95%
Catalog # APOSOR942429-250MG
Supplier:  Apollo Scientific
CAS Number:  57295-30-4
3-Hydroxy-4-methyl-benzaldehyde 95%

Bottle, 250 mg

CAS Number:  57295-30-4
Price
49,20 €
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  3-Hydroxy-4-methylbenzaldehyde
  • CAS Number:57295-30-4
  • Formula:C₈H₈O₂
  • MDL Number:MFCD01632496
  • Molecular Weight:136.148 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg

Specifications