Order Entry
Italy
Orders LinkContactUsLinkComponent
3-Hydroxy-4-methyl-benzaldehyde 95%

3-Hydroxy-4-methyl-benzaldehyde 95%

Bottle, 1 g
Catalog # APOSOR942429-1G
Supplier:   Apollo Scientific
3-Hydroxy-4-methyl-benzaldehyde 95%
Catalog # APOSOR942429-1G
Supplier:  Apollo Scientific
CAS Number:  57295-30-4
3-Hydroxy-4-methyl-benzaldehyde 95%

Bottle, 1 g

CAS Number:  57295-30-4
Price
105,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  3-Hydroxy-4-methylbenzaldehyde
  • CAS Number:57295-30-4
  • Formula:C₈H₈O₂
  • MDL Number:MFCD01632496
  • Molecular Weight:136.148 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.