Methyl-4-hydroxybenzoate 98%
Bottle, 500 g
Catalog # APOSOR922886-500G
Supplier: Apollo Scientific
Methyl-4-hydroxybenzoate 98%
Catalog # APOSOR922886-500G
Supplier: Apollo Scientific
CAS Number: 99-76-3
Jump to:
Product Details & Documents
Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
- CAS Number:99-76-3
- Formula:HOC₆H₄CO₂CH₃
- MDL Number:MFCD00002352
- Molecular Weight:152.15 g/mol
- Melting Pt:125…128 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:500 g



