Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Methyl-4-hydroxybenzoate 98%

Bottle, 1 kg
Catalog # APOSOR922886-1KG
Supplier:   Apollo Scientific
Methyl-4-hydroxybenzoate 98%
Catalog # APOSOR922886-1KG
Supplier:  Apollo Scientific
CAS Number:  99-76-3
Methyl-4-hydroxybenzoate 98%

Bottle, 1 kg

CAS Number:  99-76-3
Price
88,30 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
  • CAS Number:99-76-3
  • Formula:HOC₆H₄CO₂CH₃
  • MDL Number:MFCD00002352
  • Molecular Weight:152.15 g/mol
  • Melting Pt:125…128 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications