Order Entry
Italy
Orders LinkContactUsLinkComponent
3,4-Dichlorothiobenzamide

3,4-Dichlorothiobenzamide

Bottle, 5 g
Codice catalogo APOSOR49004-5G
Nome del fornitore:   Apollo Scientific
3,4-Dichlorothiobenzamide
Codice catalogo APOSOR49004-5G
Nome del fornitore:  Apollo Scientific
Numero CAS:  22179-73-3
3,4-Dichlorothiobenzamide

Bottle, 5 g

Numero CAS:  22179-73-3
Price
319,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:22179-73-3
  • Formula:C₇H₅Cl₂NS
  • MDL Number:MFCD04973321
  • Molecular Weight:206.09 g/mol
  • Melting Pt:133…137 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.