Order Entry
Italy
Orders LinkContactUsLinkComponent
 

3,4-Dichlorothiobenzamide

Bottle, 10 g
Catalog # APOSOR49004-10G
Supplier:   Apollo Scientific
3,4-Dichlorothiobenzamide
Catalog # APOSOR49004-10G
Supplier:  Apollo Scientific
CAS Number:  22179-73-3
3,4-Dichlorothiobenzamide

Bottle, 10 g

CAS Number:  22179-73-3
Price
521,00 €
Log in for availability
1 * 10 g
Product Details & Documents
  • CAS Number:22179-73-3
  • Formula:C₇H₅Cl₂NS
  • MDL Number:MFCD04973321
  • Molecular Weight:206.09 g/mol
  • Melting Pt:133…137 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications