Order Entry
Italy
Orders LinkContactUsLinkComponent
1,6-Hexanedithiol 97+%

1,6-Hexanedithiol 97+%

Bottle, 25 g
Catalog # APOSOR451319-25G
Irritant
Supplier:   Apollo Scientific
1,6-Hexanedithiol 97+%
Catalog # APOSOR451319-25G
Supplier:  Apollo Scientific
CAS Number:  1191-43-1
1,6-Hexanedithiol 97+%

Bottle, 25 g

CAS Number:  1191-43-1
Price
29,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1,6-Dimercaptohexane
  • CAS Number:1191-43-1
  • Formula:C₆H₁₄S₂
  • MDL Number:MFCD00004910
  • Molecular Weight:150.31 g/mol
  • Boiling Pt:118…119 °C
  • Melting Pt:–21 °C
  • Density:0.995 g/cm³ (25 °C)
  • Flash Pt:91 °C
  • Storage Temperature:Ambient
  • UN:3334
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.