Order Entry
Italy
Orders LinkContactUsLinkComponent
2,6-Dimethoxynaphthalene ≥98%

2,6-Dimethoxynaphthalene ≥98%

Bottle, 5 g
Catalog # APOSOR925404-5G
Supplier:   Apollo Scientific
2,6-Dimethoxynaphthalene ≥98%
Catalog # APOSOR925404-5G
Supplier:  Apollo Scientific
CAS Number:  5486-55-5
2,6-Dimethoxynaphthalene ≥98%

Bottle, 5 g

CAS Number:  5486-55-5
Price
39,10 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:5486-55-5
  • Formula:C₁₂H₁₂O₂
  • MDL Number:MFCD00004063
  • Molecular Weight:188.23 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.