Order Entry
Italy
Orders LinkContactUsLinkComponent
 

(4-Chlorophenyl)methyl acetate, tech

Bottle, 1 g
Catalog # APOSOR32624-1G
Supplier:   Apollo Scientific
(4-Chlorophenyl)methyl acetate, tech
Catalog # APOSOR32624-1G
Supplier:  Apollo Scientific
CAS Number:  5406-33-7
(4-Chlorophenyl)methyl acetate, tech

Bottle, 1 g

CAS Number:  5406-33-7
Price
1 010,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  p-Chlorobenzyl acetate, 4-Chlorobenzyl acetate
  • CAS Number:5406-33-7
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD00027406
  • Molecular Weight:184.62 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.