Order Entry
Italy
Orders LinkContactUsLinkComponent
6-Methoxy-2-naphthaldehyde 98%

6-Methoxy-2-naphthaldehyde 98%

Bottle, 25 g
Codice catalogo APOSOR900156-25G
Nome del fornitore:   Apollo Scientific
6-Methoxy-2-naphthaldehyde 98%
Codice catalogo APOSOR900156-25G
Nome del fornitore:  Apollo Scientific
Numero CAS:  3453-33-6
6-Methoxy-2-naphthaldehyde 98%

Bottle, 25 g

Numero CAS:  3453-33-6
Price
102,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:3453-33-6
  • Formula:C₁₂H₁₀O₂
  • MDL Number:MFCD00081152
  • Molecular Weight:186.21 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.