Order Entry
Italy
Orders LinkContactUsLinkComponent
6-Methoxy-2-naphthaldehyde 98%

6-Methoxy-2-naphthaldehyde 98%

Bottle, 100 g
Catalog # APOSOR900156-100G
Supplier:   Apollo Scientific
6-Methoxy-2-naphthaldehyde 98%
Catalog # APOSOR900156-100G
Supplier:  Apollo Scientific
CAS Number:  3453-33-6
6-Methoxy-2-naphthaldehyde 98%

Bottle, 100 g

CAS Number:  3453-33-6
Price
174,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:3453-33-6
  • Formula:C₁₂H₁₀O₂
  • MDL Number:MFCD00081152
  • Molecular Weight:186.21 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.