Order Entry
Italy
Orders LinkContactUsLinkComponent
Phenol solution 5% (w/v)

Phenol solution 5% (w/v)

500 mL
Catalog # SPCMP-259-500ML
Health
Corrosive
Supplier:   Spectrum Chemical
Phenol solution 5% (w/v)
Catalog # SPCMP-259-500ML
Supplier:  Spectrum Chemical
CAS Number:  108-95-2
Phenol solution 5% (w/v)

500 mL

CAS Number:  108-95-2
Price
64,60 €
Log in for availability
1 * 500 mL
Product Details & Documents

Phenol, 5 Percent (w/v) Solution - Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
Specifications

Specification Test Results

  • Assay (w/v):
    4.9-5.1 %

Specifications

  • Pk:
    500 mL

Specifications