Order Entry
Italy
Orders LinkContactUsLinkComponent
Phenol solution 5% (w/v)

Phenol solution 5% (w/v)

Plastic, 1 L
Catalog # SPCMP-259-1LTPL
Health
Corrosive
Supplier:   Spectrum Chemical
Phenol solution 5% (w/v)
Catalog # SPCMP-259-1LTPL
Supplier:  Spectrum Chemical
CAS Number:  108-95-2
Phenol solution 5% (w/v)

Plastic, 1 L

CAS Number:  108-95-2
Price
85,70 €
Log in for availability
1 * 1 L
Product Details & Documents

Phenol, 5 Percent (w/v) Solution - Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
Specifications

Specification Test Results

  • Assay (w/v):
    4.9-5.1 %

Specifications

  • Pack type:
    Plastic
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.