Order Entry
Italy
Orders LinkContactUsLinkComponent
Condensers for REVOLVE 3M microscope
Condensers for REVOLVE 3M microscope
ELWD universal condenser, WD 73 mm, NA 0,30 aperture
Catalog # 630-3323
Supplier:   DISCOVER ECHO
Condensers for REVOLVE 3M microscope
Catalog # 630-3323
Supplier:  DISCOVER ECHO
Condensers for REVOLVE 3M microscope
ELWD universal condenser, WD 73 mm, NA 0,30 aperture
Price
1 550,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Documents
Specifications
  • Description:
    ELWD universal condenser, WD 73 mm, NA 0,30 aperture
  • Seq:
    01