Order Entry
Italy
Orders LinkContactUsLinkComponent
1-Hexene for synthesis

1-Hexene for synthesis

135 kg
Catalog # 8.20639.9135
Flammable
Health
Supplier:   Merck
1-Hexene for synthesis
Catalog # 8.20639.9135
Supplier:  Merck
CAS Number:  592-41-6
1-Hexene for synthesis

135 kg

CAS Number:  592-41-6
Price
Inquire For Price
Log in for availability
1 * 135 kg
Product Details & Documents
  • CAS Number:592-41-6
  • Formula:C₆H₁₂
  • MDL Number:MFCD00009505
  • Molecular Weight:84.16 g/mol
  • Boiling Pt:63 °C (1013 hPa)
  • Melting Pt:–140 °C
  • Density:0.675 g/cm³ (20 °C)
  • Flash Pt:–26 °C
  • Storage Temperature:Ambient
  • EINECS:209-753-1
  • UN:2370
  • ADR:3,II

Documents

Specifications

Specifications

  • Pk:
    135 kg

Specifications