Glove R068 R-Flex seamless HPPE knit featuring Nitrile Full Dip with cut protection, high dexterity and liquid resistance. Touch Screen compatible on first two fingers and thumb. Size 9 1 * 12 PAIR
Catalog # ANSER068090
Supplier: Ansell
Frequently Bought Together