Order Entry
Italy
ContactUsLinkComponent
ScienceLine plus pH combination electrode with plug head, glass shaft, Pt junction, electrolyte KCl 3 mol/l, Silamid®-reference system with silver ion barrier and double-electrolyte, sphere membrane, H-glass, length 170mm, 12 mm Ø, -5...+100 °C, 0... 1 * 1 items
Codice catalogo SCOI285101305
Supplier:  SI Analytics
ScienceLine plus pH combination electrode with plug head, glass shaft, Pt junction, electrolyte KCl 3 mol/l, Silamid®-reference system with silver ion barrier and double-electrolyte, sphere membrane, H-glass, length 170mm, 12 mm Ø, -5...+100 °C, 0... 1 * 1 items
Codice catalogo SCOI285101305
Supplier:  SI Analytics
Frequently Bought Together