Order Entry
Italy
ContactUsLinkComponent
PERKINELMER INTRODUCES THE NEXION 5000 ICP-MS STANDARD SOLUTION-READY SAMPLE INTRODUCTION TECHNOLOGY. IT IS ENGINEERED FOR THE WIDES T RANGE OF ICP-MS ANALYSES. THIS KIT INCLUDES: FIXED 2.0 MM INJECTOR UHP QUARTZ TORCH, QTY. 2 (N8152428) SILQ CYCLONI 1 * 1 items
Catalog # PERCN8160027
Supplier:  PerkinElmer
PERKINELMER INTRODUCES THE NEXION 5000 ICP-MS STANDARD SOLUTION-READY SAMPLE INTRODUCTION TECHNOLOGY. IT IS ENGINEERED FOR THE WIDES T RANGE OF ICP-MS ANALYSES. THIS KIT INCLUDES: FIXED 2.0 MM INJECTOR UHP QUARTZ TORCH, QTY. 2 (N8152428) SILQ CYCLONI 1 * 1 items
Catalog # PERCN8160027
Supplier:  PerkinElmer
Frequently Bought Together