Order Entry
Italy
ContactUsLinkComponent
THE CONTENTS OF THIS KIT ARE DESCRIBED AS FOLLOWS: 1.0 MM ID CARBON FIBER SUPPORTED SAMPLE PROBE AQUEOUS 6-PORT VALVE VALVE PUMP TUB ING KIT NUT AND FERRULE PLACEMENT FITTING (10) CTFE FERRULES, 1/16 INTERNAL STANDARD TEE KIT PASSIVE BUBBLER TEE PEE 1 * 1 items
Catalog # PERCN0810949
Supplier:  PerkinElmer
THE CONTENTS OF THIS KIT ARE DESCRIBED AS FOLLOWS: 1.0 MM ID CARBON FIBER SUPPORTED SAMPLE PROBE AQUEOUS 6-PORT VALVE VALVE PUMP TUB ING KIT NUT AND FERRULE PLACEMENT FITTING (10) CTFE FERRULES, 1/16 INTERNAL STANDARD TEE KIT PASSIVE BUBBLER TEE PEE 1 * 1 items
Catalog # PERCN0810949
Supplier:  PerkinElmer
Frequently Bought Together