Order Entry
Italy
ContactUsLinkComponent
Volumetric flask, 250 ml, class A, conformity certified, ST 29/32, PE-stopper, Diffico brown, made from DURAN tubing, All volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer 1 * 10 items
Codice catalogo WITG3.664.025.29
Nome del fornitore:  witeg Labortechnik
Volumetric flask, 250 ml, class A, conformity certified, ST 29/32, PE-stopper, Diffico brown, made from DURAN tubing, All volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer 1 * 10 items
Codice catalogo WITG3.664.025.29
Nome del fornitore:  witeg Labortechnik
Frequently Bought Together