AriaMx real-time PCR system
Product Details & Documents
The AriaMx real-time PCR system is a fully integrated quantitative PCR amplification, detection and data analysis system.
- Gene expression analysis
- New genotyping/HRM capability, mRNA quantification
- NGS quantification: Library preparation, result validation, nucleic acid monitoring and rare allele detection
- Can use self-installable optical cartridges
- Operates at a Cq uniformity standard deviation of <0,20 at fast cycling (5 s at 95 °C/10 s at 60 °C)
- Data acquisition time of <3 s/plate
- Dynamic range of nine orders of magnitude
- Operates at a maximum altitude of 2000 m and in 20 to 80% relative humidity
The system design combines a state-of-the-art thermal cycler, an advanced optical system with an LED excitation source, and complete data analysis software.
This instrument can hold up to six optics modules, and the scanning optics design delivers optimal separation between the dyes and between samples also providing a 'closed tube' PCR detection format that can be used with a variety of fluorescence detection chemistries including SYBR® Green and EvaGreen dyes, as well as fluorogenic probe systems including TaqMan probes.
The AriaMx amplifies the productivity with its unique modular and flexible design, intuitive touch screen interface, advanced, easy to use reporting and 120+ attributes monitored through the built-in on-board diagnostics to help pinpoint assay or instrument issues as they arise.
: Software upgrade statements available online, no charge. Optional for purchase warranty length of five years.
Includes configurable optical modules (up to six), a touch screen controller, Experimental Import, LIMS connectivity, 96 wells, 5 GB of protocol storage, on-board instrument diagnostics, remote monitoring capabilities, one year standard warranty.
: For research use only. Not for use in diagnostic procedures.
Documents
Agilent AriaMx Real-Time PCR System
Product Literature
Specifications
- Description:AriaMx Electronic Tracking Software
- Seq:10
- Details:21 CFR 11 Compliance software



